Merck
  • Platinum nanoparticles on porphyrin functionalized graphene nanosheets as a superior catalyst for methanol electrooxidation.
Platinum nanoparticles on porphyrin functionalized graphene nanosheets as a superior catalyst for methanol electrooxidation.
Nanoscale (2014-11-05)
Rui-Xiang Wang, Jing-Jing Fan, You-Jun Fan, Jing-Ping Zhong, Li Wang, Shi-Gang Sun, Xing-Can Shen
ABSTRACT
A novel nanostructured catalyst of platinum nanoparticles supported on 5,10,15,20-tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate) (TMPyP) functionalized graphene (TMPyP-graphene) is synthesized by the hydrothermal polyol process. The as-synthesized nanocomposites are characterized by Fourier transform infrared (FTIR) spectroscopy, UV-vis absorption spectroscopy, Raman spectroscopy, X-ray diffraction (XRD), transmission electron microscopy (TEM), X-ray photoelectron spectroscopy (XPS) and electrochemical tests. It has been found that Pt nanoparticles of ca. 3.4 nm are uniformly dispersed on the surface of TMPyP-graphene, and hold a high electrochemical active surface area (ECSA) of 126.2 m(2) g(-1). The results demonstrate that the Pt/TMPyP-graphene catalyst exhibits a much higher electrocatalytic activity and stability than the Pt/graphene and commercial Pt/C catalysts for methanol oxidation, which is of significant importance in improving the efficiency of Pt-based electrocatalysts for DMFCs applications.
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroplatinic acid hexahydrate, BioXtra
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroplatinic acid hexahydrate, ACS reagent, ≥37.50% Pt basis
Sigma-Aldrich
Methanol, Absolute - Acetone free