Merck
  • Particle size distribution and column efficiency. An ongoing debate revived with 1.9μm Titan-C18 particles.
Particle size distribution and column efficiency. An ongoing debate revived with 1.9μm Titan-C18 particles.
Journal of chromatography. A (2014-07-09)
Fabrice Gritti, David S Bell, Georges Guiochon
ABSTRACT
The mass transfer mechanism in four prototype columns (2.1 and 3.0×50mm, 2.1 and 3.0×100mm) packed with 1.9μm fully porous Titan-C18 particles was investigated by using two previously reported home-made protocols. The first one was used to measure the eddy dispersion HETP of these new columns, the second one to estimate their intrinsic (corrected for HPLC system contribution) HETPs. Titan particles are fully porous particles with a narrow particle size distribution (RSD of 9.2%). The mean Sauter diameter (dSauter=2.04μm) was determined from Coulter counter measurements on the raw silica material (before C18 derivatization) and in the absence of a dispersant agent (Triton X-100) in a 2% NaCl electrolyte solution. The results show that these RPLC Titan columns have intrinsic minimum reduced HETPs ranging from 1.7 to 1.9 and generate up to 290,000 plates per meter. The 3.0mm i.d. columns are more efficient than the 2.1mm i.d. ones and short columns are preferred to minimize efficiency losses due to frictional heating at high speeds. This work also revealed that (1) the lowest h values of the Titan columns are observed at low reduced velocities (νopt=5); (2) this is due to the unusually small diffusivity of analytes across the porous Titan-C18 particles; and (3) the Titan columns are not packed more uniformly than conventional columns packed with fully porous particles. Earlier and recent findings showing that the PSD has no direct physical impact on eddy dispersion and column efficiency are confirmed by these results.
Sigma-Aldrich
Valerophenone, 99%
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Acetanilide, purified by sublimation, ≥99.9%
Sigma-Aldrich
Acetanilide, zone-refined, purified by sublimation, ≥99.95%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Acetanilide, 99%
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetanilide, puriss. p.a., ≥99.5% (CHN)
Sigma-Aldrich
Acetophenone, natural, 98%, FG
Supelco
Acetophenone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acetanilide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetanilide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Octanophenone, ≥99%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Acetaminophen Related Compound D, Pharmaceutical Secondary Standard; Certified Reference Material