Skip to Content
Merck
All Photos(1)

Documents

V659

Sigma-Aldrich

Valerophenone

99%

Synonym(s):

Butyl phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO(CH2)3CH3
CAS Number:
Molecular Weight:
162.23
Beilstein:
1907717
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.5143 (lit.)

bp

244-245 °C (lit.)

density

0.975 g/mL at 20 °C (lit.)

SMILES string

CCCCC(=O)c1ccccc1

InChI

1S/C11H14O/c1-2-3-9-11(12)10-7-5-4-6-8-10/h4-8H,2-3,9H2,1H3

InChI key

XKGLSKVNOSHTAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valerophenone is a prochiral ketone which can undergo Norrish Type II reaction and hence can be used as a UV actinometer in photochemical experiments.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fabrice Gritti
Journal of chromatography. A, 1597, 119-131 (2019-03-25)
Three different approaches designed to transfer gradient methods from a chromatographic column 1 packed with particles (2.7 μm 90 ÅCORTECS-C18 or CORTECS-Triphenyl) to a column 2 packed with the same particles having a larger average pore diameter (APD = 120 Åand 450 Å) are proposed for
Photochemistry of valerophenone in solid solutions
Klan P, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 134(1-2), 37-44 (2000)
Dmitry I Osolodkin et al.
ACS medicinal chemistry letters, 4(9), 869-874 (2014-06-06)
Flaviviruses form a large family of enveloped viruses affecting millions of people over the world. To date, no specific therapy was suggested for the infected people, making the treatment exclusively symptomatic. Several attempts were performed earlier for the design of
Fabrice Gritti et al.
Journal of chromatography. A, 1593, 34-46 (2019-02-02)
The chromatographic performance (speed, efficiency, and gradient peak capacity for the same analysis time) of conical columns are investigated from fundamental and experimental viewpoints. A stainless steel, conically shaped column (2.1 mm i.d/4.2 mm i.d. × 15 cm long, 0.4°
Martin Gilar et al.
Journal of chromatography. A, 1523, 215-223 (2017-07-25)
The speed and separation performance of high-throughput liquid chromatography (HT LC) was investigated. We evaluated the contributions of various experimental parameters to the total analysis cycle time, including column length (column void time), gradient delay, flow rate, auto-sampler (A/S) speed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service