Merck
  • Synthesis and characterization of carbon nanofibers by catalytic chemical vapor deposition using non-ferromagnetic metal complexes.
Synthesis and characterization of carbon nanofibers by catalytic chemical vapor deposition using non-ferromagnetic metal complexes.
Journal of nanoscience and nanotechnology (2014-04-18)
Chi-Jung Su, Wei-Li Yuan, Tzu-Wei Lai, Chien-Ming Lei
ABSTRACT
Carbon nanofibers (CNFs) have wide applications in energy storage devices, electrically conducting composites, selective adsorbents, and catalyst supports. Catalytic chemical vapor deposition was carried out in this work to synthesize CNFs at mild temperatures of 700 and 800 degrees C. Non-ferromagnetic metal complexes of La, Nb, and Ti, spread on porous NaX-type zeolite support, were tested as new catalyst. CNFs ranging from 30 to 200 nm in diameter were obtained. Images of transmission electron microscopy showed encapsulated transition-metal nanoparticles by CNFs. X-ray diffraction patterns revealed the crystalline structures of La (FCC), Nb (BCC), and Ti (HCP) formed over zeolite. Magnetic hysteresis loops showed superconductivity from the CNF-encapsulated Nb at 2 K. Raman spectra showed that all the samples possessed graphitic and amorphous carbon structures. Based on the SEM images and Raman spectra, the three metals all catalyzed the synthesis of CNFs.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, puriss. p.a., powder
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon, foil, 100x100mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon