Skip to Content
Merck
All Photos(1)

Key Documents

37771

Sigma-Aldrich

Activated Charcoal Norit®

greener alternative

Norit® GAC 1240W, from coal, steam activated, granular

Synonym(s):

Charcoal activated Norit®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Beilstein:
4360473
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

form

granular

quality

steam activated

manufacturer/tradename

Norit® GAC 1240W

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

ion chromatography: suitable

diam.

~1 mm

ign. residue

≤12%

loss

≤5.0% loss on drying

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated Charcoal Norit® (Norit GAC 1240W) can be used in the extraction of polycyclic aromatic hydrocarbons (PAHs) and hydrophobic organic contaminants from highly contaminated sediments.[1][2]

Legal Information

Norit is a registered trademark of Norit N.V.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction of sediment-associated polycyclic aromatic hydrocarbons with granular activated carbon
Rakowska MI, et al.
Environmental Toxicology and Chemistry / Setac, 32(2), 304-311 (2013)
Kinetics of hydrophobic organic contaminant extraction from sediment by granular activated carbon
Rakowska MI, et al.
Water Research, 51(2), 86-95 (2014)
Performance of carbon nanomaterial (nanotubes and nanofibres) supported platinum and palladium catalysts for the hydrogenation of cinnamaldehyde and of 1-octyne.
Jung A, et al.
Applied Catalysis A: General, 362(1), 95-105 (2009)
Use of activated charcoal for the removal of patulin from cider.
Sands DC, et al.
Applied and Environmental Microbiology, 32(3), 388-391 (1976)
Diazepam stability in wastewater and removal by advanced membranes technology, activated carbon and micelle-clay complex
Bufo SA, et al.
Desalination and Water Treatment (2015)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service