Merck
  • Copper-catalyzed α-methylenation of benzylpyridines using dimethylacetamide as one-carbon source.
Copper-catalyzed α-methylenation of benzylpyridines using dimethylacetamide as one-carbon source.
Organic letters (2014-03-22)
Masaki Itoh, Koji Hirano, Tetsuya Satoh, Masahiro Miura
ABSTRACT
The direct α-methylenation of benzylpyridines was achieved using N,N-dimethylacetamide (DMA) as a one-carbon source under copper catalysis. An intermediary species was detected at an early stage, and a possible mechanism was proposed. Additionally, α-oxygenation and dimerization of benzylpyridines could also be performed efficiently.
Product Description
USP
Residual Solvent Class 2 - N,N-Dimethylacetamide, United States Pharmacopeia (USP) Reference Standard
Pheniramine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Benzylpyridine, 98%
Sigma-Aldrich
N,N-Dimethylacetamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylacetamide, SAJ special grade
Supelco
N,N-Dimethylacetamide, analytical standard
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylacetamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylacetamide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for HPLC, ≥99.9%