Skip to Content
Merck
All Photos(1)

Key Documents

07-4072

Sigma-Aldrich

N,N-Dimethylacetamide

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

grade

SAJ first grade

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

availability

available only in Japan

refractive index

n20/D 1.437 (lit.)

pH

4 (20 °C, 200 g/L)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

145.4 °F

Flash Point(C)

63 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sood et al.
Animal reproduction science, 129(1-2), 89-95 (2011-11-15)
Three experiments conducted to improve the survival of emu sperm during cryopreservation aimed to: (1) minimize chilling injury during the cooling phase; (2) determine the osmotic effects of dimethylacetamide (DMA), sucrose and trehalose; and (3) investigate the timing and nature
Tony Reid et al.
The Lancet. Oncology, 16(9), 1133-1142 (2015-08-25)
Epigenetic alterations have been strongly associated with tumour formation and resistance to chemotherapeutic drugs, and epigenetic modifications are an attractive target in cancer research. RRx-001 is activated by hypoxia and induces the generation of reactive oxygen and nitrogen species that
Jingshuai Yang et al.
ChemSusChem, 6(2), 275-282 (2013-01-11)
Covalently cross-linked polymer membranes were fabricated from poly(aryl sulfone benzimidazole) (SO(2)PBI) and poly(vinylbenzyl chloride) (PVBCl) as electrolytes for high-temperature proton-exchange-membrane fuel cells. The cross-linking imparted organo insolubility and chemical stability against radical attack to the otherwise flexible SO(2)PBI membranes. Steady
Wesley Walker et al.
Journal of the American Chemical Society, 135(6), 2076-2079 (2013-01-31)
A major challenge in the development of rechargeable Li-O(2) batteries is the identification of electrolyte materials that are stable in the operating environment of the O(2) electrode. Straight-chain alkyl amides are one of the few classes of polar, aprotic solvents
Fen Ran et al.
Acta biomaterialia, 7(9), 3370-3381 (2011-06-11)
An amphiphilic triblock co-polymer of poly(vinyl pyrrolidone)-b-poly(methyl methacrylate)-b-poly(vinyl pyrrolidone) (PVP-b-PMMA-b-PVP) was synthesized via reversible addition-fragmentation chain transfer (RAFT) polymerization. The block co-polymer can be directly blended with polyethersulfone (PES) using dimethylacetamide (DMAC) as the solvent to prepare flat sheet and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service