Merck
  • Intraperitoneal chemotherapy for peritoneal surface malignancy: experience with 1,000 patients.
Intraperitoneal chemotherapy for peritoneal surface malignancy: experience with 1,000 patients.
Journal of the American College of Surgeons (2014-02-05)
Edward A Levine, John H Stewart, Perry Shen, Gregory B Russell, Brian L Loggie, Konstantinos I Votanopoulos
ABSTRACT
Peritoneal dissemination of abdominal malignancy (carcinomatosis) has a clinical course marked by bowel obstruction and death; it traditionally does not respond well to systemic therapy and has been approached with nihilism. To treat carcinomatosis, we use cytoreductive surgery (CS) with hyperthermic intraperitoneal chemotherapy (HIPEC). A prospective database of patients has been maintained since 1992. Patients with biopsy-proven peritoneal surface disease were uniformly evaluated for, and treated with, CS and HIPEC. Patient demographics, performance status (Eastern Cooperative Oncology Group), resection status, and peritoneal surface disease were classified according to primary site. Univariate and multivariate analyses were performed. The experience was divided into quintiles and outcomes compared. Between 1991 and 2013, a total of 1,000 patients underwent 1,097 HIPEC procedures. Mean age was 52.9 years and 53.1% were female. Primary tumor site was appendix in 472 (47.2%), colorectal in 248 (24.8%), mesothelioma in 72 (7.2%), ovary in 69 (6.9%), gastric in 46 (4.6%), and other in 97 (9.7%). Thirty-day mortality rate was 3.8% and median hospital stay was 8 days. Median overall survival was 29.4 months, with a 5-year survival rate of 32.5%. Factors correlating with improved survival on univariate and multivariate analysis (p ≤ 0.0001 for each) were preoperative performance status, primary tumor type, resection status, and experience quintile (p = 0.04). For the 5 quintiles, the 1- and 5-year survival rates, as well as the complete cytoreduction score (R0, R1, R2a) have increased, and transfusions, stoma creations, and complications have all decreased significantly (p < .001 for all). This largest reported single-center experience with CS and HIPEC demonstrates that prognostic factors include primary site, performance status, completeness of resection, and institutional experience. The data show that outcomes have improved over time, with more complete cytoreduction and fewer serious complications, transfusions, and stomas. This was due to better patient selection and increased operative experience. Cytoreductive surgery with HIPEC represents a substantial improvement in outcomes compared with historical series, and shows that meaningful long-term survival is possible for selected carcinomatosis patients. Multi-institutional cooperative trials are needed to refine the use of CS and HIPEC.
Sigma-Aldrich
Sodium thiosulfate solution, 0.025 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.1 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.01 M
Sigma-Aldrich
Sodium thiosulfate solution, 1 M
Sigma-Aldrich
Sodium thiosulfate pentahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Sigma-Aldrich
Sodium thiosulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium thiosulfate pentahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP
Mitomycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
Carboplatin
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%