Skip to Content
Merck
All Photos(1)

Key Documents

Y0000271

Oxaliplatin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

[SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N′)[ethanedioata(2--)-O,O’]platinum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O4Pt
CAS Number:
Molecular Weight:
397.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxaliplatin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1O[Pt]OC1=O.N[C@@H]2CCCC[C@H]2N

InChI

1S/C6H14N2.C2H2O4.Pt/c7-5-3-1-2-4-6(5)8;3-1(4)2(5)6;/h5-6H,1-4,7-8H2;(H,3,4)(H,5,6);/q;;+2/p-2/t5-,6-;;/m1../s1

InChI key

ZROHGHOFXNOHSO-BNTLRKBRSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxaliplatin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Lact. - Muta. 2 - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M E Engelen et al.
European journal of cancer (Oxford, England : 1990), 49(10), 2311-2320 (2013-04-11)
The purpose of this multicenter cohort study was to evaluate whether a differentiated treatment of primary rectal cancer based on magnetic resonance imaging (MRI) can reduce the number of incomplete resections and local recurrences and improve recurrence-free and overall survival.
Roser Velasco et al.
Journal of neurology, neurosurgery, and psychiatry, 85(4), 392-398 (2013-07-03)
Peripheral neuropathy ranks among the most common dose-limiting and disabling side-effect of oxaliplatin (OXA)-based chemotherapy. The aim of this prospective, multicentre study was to define early clinical and neurophysiological markers that may help to identify patients at risk of developing
Catia Moutinho et al.
Journal of the National Cancer Institute, 106(1), djt322-djt322 (2013-11-26)
A major problem in cancer chemotherapy is the existence of primary resistance and/or the acquisition of secondary resistance. Many cellular defects contribute to chemoresistance, but epigenetic changes can also be a cause. A DNA methylation microarray was used to identify
Bernard Nordlinger et al.
The Lancet. Oncology, 14(12), 1208-1215 (2013-10-15)
Previous results of the EORTC intergroup trial 40983 showed that perioperative chemotherapy with FOLFOX4 (folinic acid, fluorouracil, and oxaliplatin) increases progression-free survival (PFS) compared with surgery alone for patients with initially resectable liver metastases from colorectal cancer. Here we present
Noriho Iida et al.
Science (New York, N.Y.), 342(6161), 967-970 (2013-11-23)
The gut microbiota influences both local and systemic inflammation. Inflammation contributes to development, progression, and treatment of cancer, but it remains unclear whether commensal bacteria affect inflammation in the sterile tumor microenvironment. Here, we show that disruption of the microbiota

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service