Merck
  • Rat pancreatitis produced by 13-week administration of zinc oxide nanoparticles: biopersistence of nanoparticles and possible solutions.
Rat pancreatitis produced by 13-week administration of zinc oxide nanoparticles: biopersistence of nanoparticles and possible solutions.
Journal of applied toxicology : JAT (2013-02-15)
Seung Hyeok Seok, Wan-Seob Cho, Jung Shin Park, Yirang Na, Ahram Jang, Hojoong Kim, Yujin Cho, Taesung Kim, Ji-Ran You, Sanghoon Ko, Byeong-Cheol Kang, Jong Kwon Lee, Jayoung Jeong, Jeong-Hwan Che
ABSTRACT
Zinc oxide (ZnO) nanoparticles (NPs) are used in diverse applications ranging from paints and cosmetics to biomedicine and food. Although micron-sized ZnO is a traditional food supplement, ZnO NPs are an unknown public health risk because of their unique physicochemical properties. Herein, we studied the 13-week subchronic toxicity of ZnO NPs administered via the oral route according to Organization for Economic Cooperation and Development (OECD) test guideline 408. Well-dispersed ZnO NPs were administered to Sprague-Dawley (SD) rats (11/sex/group) at doses of 67.1, 134.2, 268.4 or 536.8 mg kg(-1) per body weight over a 13-week period. The mean body weight gain in males given 536.8 mg kg(-1) ZnO NPs was significantly lower than that of control male rats, whereas no significant differences were observed between the other treatment groups and the controls. Male and female rats dosed at 536.8 mg kg(-1) ZnO NPs had significant changes in anemia-related hematologic parameters. Mild to moderate pancreatitis also developed in both sexes dosed at 536.8 mg kg(-1) , whereas no histological changes were observed in the other treatment groups. To evaluate the mechanism of toxicity, we performed a bio-persistence study and evaluated the effects of the ZnO NPs on cell proliferation. The treatment of a human gastric adenocarcinoma cell line with ZnO NPs resulted in a significant inhibition of cellular proliferation. The anti-proliferative effect of ZnO NPs or Zn(2+) was effectively blocked by treatment with chelators. These results indicate that the bio-persistence of ZnO NPs after ingestion is key to their toxicity; the no-observed-adverse effect level (NOAEL) of ZnO NPs was found to be 268.4 mg kg(-1) per day for both sexes.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, SAJ first grade, ≥98.0%
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.02 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.1 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.05 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.01 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution