Skip to Content
Merck
All Photos(1)

Key Documents

2854

Sigma-Aldrich

Ethylenediaminetetraacetic acid disodium salt solution

2%, solution

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid disodium salt, EDTA disodium salt, EDTA-Na2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2N(CH2CO2Na)CH2CO2H]2
CAS Number:
Molecular Weight:
336.21
Beilstein:
3822669
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

solution

Quality Level

concentration

2%

storage temp.

2-8°C

SMILES string

[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;2*+1/p-2

InChI key

ZGTMUACCHSMWAC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediamine tetraacetate (EDTA) is a calcium ion chelator,[1] that has a low molecular mass of 292.24 Da.[2]

Application


  • Dual-Responsive Hydrogels for Mercury Ion Detection and Removal from Wastewater.: This study discusses the application of ethylenediaminetetraacetic acid disodium salt in the development of dual-responsive hydrogels, highlighting its effectiveness in detecting and removing mercury ions from wastewater (Diacon et al., 2024).

  • Photocatalytic Hydrogen Evolution Activity of Nitrogen/Fluorine-Codoped Rutile TiO(2).: The article explores the role of ethylenediaminetetraacetic acid disodium salt in enhancing the photocatalytic activity of nitrogen/fluorine-codoped rutile TiO(2) for hydrogen evolution (Miyoshi et al., 2023).

  • High-efficiency cleaning technology and lifespan prediction for the ceramic membrane treating secondary treated effluent.: This study presents a novel cleaning technology involving ethylenediaminetetraacetic acid disodium salt for ceramic membranes, offering insights into efficiency enhancements and lifespan predictions (Lv et al., 2023).

  • Reversible Morphology Locking via Metal Infiltration in a Block Copolymer.: This research investigates the reversible morphology locking of block copolymers facilitated by metal infiltration, using ethylenediaminetetraacetic acid disodium salt as a key component in the process (Ma et al., 2023).

Biochem/physiol Actions

Ethylenediaminetetraacetic acid disodium salt has the ability to block the binding of vasoactive intestinal peptide to macrophage membranes.[1] It is mainly used in the purification of protein, to remove divalent cations and also to prevent protease activity.[2]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drawbacks of dialysis procedures for removal of EDTA
Monico A, et al.
PLoS ONE, 12(1), e0169843-e0169843 (2017)
Sabina Müller et al.
Methods in molecular biology (Clifton, N.J.), 2654, 463-476 (2023-04-28)
CD8+ cytotoxic T lymphocytes (CTL) play a key role in anti-tumor immune response. They are therefore at the heart of current immunotherapy protocols against cancer. Despite current strategies to potentiate CTL responses, cancer cells can resist CTL attack, thus limiting
J J Segura et al.
Journal of endodontics, 22(7), 337-340 (1996-07-01)
The purpose of this study was to investigate the effect of the disodium salt of ethylenediamine tetraacetate (EDTA), a calcium ion chelator used in the root canal therapy, on vasoactive intestinal peptide (VIP) binding to macrophage membranes (MM's). Binding assays
Rebecca Wüst et al.
Frontiers in bioengineering and biotechnology, 10, 964705-964705 (2022-10-11)
Vascularization of tissue-engineered constructs remains a key challenge in the field of skeletal muscle tissue engineering. One strategy for vascularizing organoids is in vitro pre-vascularization, relying on de novo assembly of undifferentiated endothelial cells into capillaries, a process termed vasculogenesis.
J L D'Amato et al.
Poultry science, 89(5), 938-947 (2010-04-08)
Arginine is an essential amino acid in Aves and is also an important substrate for the immune system. Dietary Arg in avian diets must be sufficient to not only support growth but also immunity. To better understand Arg needs for

Questions

  1. Is it acceptable to use 285-4 after it has been stored at room temperature for 14 days?

    1 answer
    1. The recommended long-term storage for the 2% EDTA solution is at 2-8°C. The product is not considered temperature-sensitive for short-term storage and, therefore, product 285-4 is shipped at ambient temperature. It is stability tested for up to 14 days at ambient temperature. The product is expected to be placed at 2-8°C within 14 days of arrival. If the solution is stored at room temperature for 7 days, it should still be suitable for use. It is advisable to check specimens anticoagulated with 285-4 for signs of clumping or clotting. If any clumping or clotting is observed, the bottle kept at room temperature should be properly disposed of, and a fresh bottle should be ordered.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service