Merck
  • Sub-micronic capsules based on gelatin and poly(maleic anhydride-alt-vinyl acetate) obtained by interfacial condensation with potential biomedical applications.
Sub-micronic capsules based on gelatin and poly(maleic anhydride-alt-vinyl acetate) obtained by interfacial condensation with potential biomedical applications.
Journal of nanoscience and nanotechnology (2013-07-19)
Delia Mihaela Iurea, Cătălina Anişoara Peptu, Jean-François Chailan, Pascal Carriere, Marcel Popa
ABSTRACT
New sub-micronic capsules based on a copolymer of maleic anhydride-alt-vinyl acetate and a natural polymer (gelatin) using an interfacial condensation method were obtained. Sub-micronic capsules were characterized by Fourier Transform infrared spectroscopy (FTIR), dynamic light scattering (DLS) method, zeta-potential, scanning electron microscopy (SEM) and atomic force microscopy (AFM). The thermal properties were investigated by thermogravimetric analysis (TGA). According to some parameters of the synthesis reaction (polymer weight ratio, acetone/water ratio, surfactant concentration), the mean diameter of the sub-micronic capsules can be tuned from 200 to 760 nm. The sub-micronic capsules show a higher agglomeration tendency as the amount of gelatin in their composition increases. The swelling capacity in aqueous solutions is dependent on the composition and size of the sub-micronic capsules, decreasing with their diameter and gelatin composition. The drug loading and release capacity was studied using Penicillin G (sodium salt) (PG), and it has been proved that it is influenced by the sub-micronic capsules morphology induced by preparation parameters. Encapsulation and controlled release of small molecule were successfully carried out, demonstrating the potential biomedical applications of these new easily obtained sub-micronic capsules.
Product Description
Supelco
Vinyl acetate solution, certified reference material, 5000 μg/mL in acetonitrile
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Supelco
Vinyl acetate, analytical standard
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Vinyl acetate, CP
Vinyl acetate, European Pharmacopoeia (EP) Reference Standard
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength