Merck
  • In vivo stimulus-induced vasodilation occurs without IP3 receptor activation and may precede astrocytic calcium increase.
In vivo stimulus-induced vasodilation occurs without IP3 receptor activation and may precede astrocytic calcium increase.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-05-10)
Krystal Nizar, Hana Uhlirova, Peifang Tian, Payam A Saisan, Qun Cheng, Lidia Reznichenko, Kimberly L Weldy, Tyler C Steed, Vishnu B Sridhar, Christopher L MacDonald, Jianxia Cui, Sergey L Gratiy, Sava Sakadzić, David A Boas, Thomas I Beka, Gaute T Einevoll, Ju Chen, Eliezer Masliah, Anders M Dale, Gabriel A Silva, Anna Devor
ABSTRACT
Calcium-dependent release of vasoactive gliotransmitters is widely assumed to trigger vasodilation associated with rapid increases in neuronal activity. Inconsistent with this hypothesis, intact stimulus-induced vasodilation was observed in inositol 1,4,5-triphosphate (IP3) type-2 receptor (R2) knock-out (KO) mice, in which the primary mechanism of astrocytic calcium increase-the release of calcium from intracellular stores following activation of an IP3-dependent pathway-is lacking. Further, our results in wild-type (WT) mice indicate that in vivo onset of astrocytic calcium increase in response to sensory stimulus could be considerably delayed relative to the simultaneously measured onset of arteriolar dilation. Delayed calcium increases in WT mice were observed in both astrocytic cell bodies and perivascular endfeet. Thus, astrocytes may not play a role in the initiation of blood flow response, at least not via calcium-dependent mechanisms. Moreover, an increase in astrocytic intracellular calcium was not required for normal vasodilation in the IP3R2-KO animals.
Product Description
Supelco
Dextran, analytical standard, for GPC, 150,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran, analytical standard, for GPC, 670,000
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Supelco
Dextran, analytical standard, for GPC, 12,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Supelco
Dextran, analytical standard, for GPC, 80,000
Supelco
Dextran, analytical standard, for GPC, 50,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
Supelco
Dextran, analytical standard, for GPC, 1,000