Skip to Content
Merck

Skip To

E3889

Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid

Molecular Biology, ≥97.0%

Synonym(s):

EGTA, Egtazic acid, Ethylene-bis(oxyethylenenitrilo)tetraacetic acid, Glycol ether diamine tetraacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
[-CH2OCH2CH2N(CH2CO2H)2]2
CAS Number:
Molecular Weight:
380.35
UNSPSC Code:
12352106
NACRES:
NA.31
PubChem Substance ID:
EC Number:
200-651-2
Beilstein/REAXYS Number:
1717370
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

assay

≥97.0%

form

powder

reaction suitability

reagent type: chelator

loss

≤1.0% loss on drying

mp

241 °C (dec.) (lit.)

foreign activity

DNAse (exonuclease), NICKase (endonuclease), RNAse, and protease, none detected

SMILES string

OC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C14H24N2O10/c17-11(18)7-15(8-12(19)20)1-3-25-5-6-26-4-2-16(9-13(21)22)10-14(23)24/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

DEFVIWRASFVYLL-UHFFFAOYSA-N

Application

A chelating agent useful for the determination of calcium in the presence of magnesium.
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid has been used:
  • in Dulbecco′s phosphate-buffered saline (DPBS) to induce the weakening of cell-to-cell junctions
  • as an extracellular calcium chelator to study the role of calcium in axonal mitochondria degeneration
  • as a component in lysis buffer for lysing mouse mucosal cells for western blot analysis

Biochem/physiol Actions

Ethylene glycol tetraacetic acid (EGTA) can chelate calcium (Ca2+) at a ratio of 1:1. It serves as an anti-coagulant when dissolved at 1 g per 100 ml of blood. EGTA can prevent the vesicular release in the nanodomain of single Ca2+ channels. This metal chelator can inhibit the activity of pseudolysin.
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid A is a chelating agent used for the determination of calcium in the presence of magnesium.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E0396037774100-OP
grade

Molecular Biology

grade

-

grade

Molecular Biology

grade

-

assay

≥97.0%

assay

≥97 .0%

assay

≥99.0% (T)

assay

-

Quality Level

400

Quality Level

400

Quality Level

300

Quality Level

100

mp

241 °C (dec.) (lit.)

mp

241 °C (dec.) (lit.)

mp

241 °C (dec.) (lit.)

mp

-

form

powder

form

powder

form

powder

form

crystalline powder (Fine)

foreign activity

DNAse (exonuclease), NICKase (endonuclease), RNAse, and protease, none detected

foreign activity

-

foreign activity

-

foreign activity

DNAse, none detected, Protease, none detected, RNAse, none detected


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Walter Marcotti et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(16), 5505-5514 (2014-04-18)
Tip links between adjacent stereocilia are believed to gate mechano-electrical transducer (MET) channels and mediate the electrical responses of sensory hair cells. We found that mouse auditory hair cells that lack tip links due to genetic mutations or exposure to
David R Whitfield et al.
The American journal of geriatric psychiatry : official journal of the American Association for Geriatric Psychiatry, 23(2), 141-148 (2014-06-24)
Depression is a common symptom in dementia with Lewy bodies (DLB), Parkinson disease dementia (PDD), and Alzheimer disease (AD), yet its molecular basis remains unclear and current antidepressants do not appear to be effective. Cerebral zinc has been implicated in
Jennifer R Brum et al.
Science (New York, N.Y.), 348(6237), 1261498-1261498 (2015-05-23)
Viruses influence ecosystems by modulating microbial population size, diversity, metabolic outputs, and gene flow. Here, we use quantitative double-stranded DNA (dsDNA) viral-fraction metagenomes (viromes) and whole viral community morphological data sets from 43 Tara Oceans expedition samples to assess viral



Global Trade Item Number

SKUGTIN
E3889-100G04061835544424
E3889-10G04061835544431
E3889-25G04061835521845
E3889-500G04061833603451

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters