Merck
  • Photoluminescence and thermoanalytical studies of complexes based on 5-Cl-8-hydroxyquinoline and calix[4]arene ligands.
Photoluminescence and thermoanalytical studies of complexes based on 5-Cl-8-hydroxyquinoline and calix[4]arene ligands.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Marcos N Soares, Elder F Moscardini, Lucildes P Mercuri, Izilda A Bagatin
ABSTRACT
A innovative 5-Cl-8-oxyquinolinepropoxycalix[4]arene ligand (2) have been prepared, exhibiting, at room temperature, blue fluorescent light emission and resulting in shift band to green fluorescent light (fluorescence mode) in the presence of coordinated Eu(III) and Tb(III) ions. Terbium complex presented phosphorescence emission as noted by typical bands at 490 nm, 545 nm and 585 nm. TG/DTG data exhibited typical thermal behavior for these compounds, however DSC curves showed the melting temperature near 300 °C for the samples, demonstrating an unusual thermal stability when quinoline derivatives are attached to calix[4]arene matrix. This fact strongly suggests an effective approach to preparing the photoluminescent compound associating high chemical and thermal stability.
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
5-Chloro-8-quinolinol, 95%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC