Merck
  • Pharmacokinetics and residues of tetracycline in crucian carp muscle using capillary electrophoresis on-line coupled with electrochemiluminescence detection.
Pharmacokinetics and residues of tetracycline in crucian carp muscle using capillary electrophoresis on-line coupled with electrochemiluminescence detection.
Food chemistry (2013-02-28)
Biyang Deng, Quanxiu Xu, Hua Lu, Li Ye, Yingzi Wang
ABSTRACT
A novel and sensitive determination for tetracycline (TC) in crucian carp muscle was developed by using capillary electrophoresis coupled with electrochemiluminescence (ECL) detection. The conditions affected separation and detection were examined in detail. The linearity range of TC concentration was from 0.005 to 10 μg mL(-1) with a correlation coefficient of 0.9992. The detection limit of TC (S/N=3) was 1.8 ng mL(-1). The relative standard deviations of the ECL intensity and the migration time for eleven consecutive injections of 1.0 μg mL(-1) TC were 1.6% and 0.8%, respectively. The recovery of TC was 97.8% (n=5). After administration of 75 mg kg(-1) TC, the maximum concentration and peak time of TC in crucian carp muscle were 7.33 mg kg(-1) and 8h, respectively. After administration of TC, TC concentration demonstrated trivial variation in the period from 48 to 96 h.
Product Description
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)