Merck
  • The fouling hydroid Ectopleura larynx: a lack of effect of next generation antifouling technologies.
The fouling hydroid Ectopleura larynx: a lack of effect of next generation antifouling technologies.
Biofouling (2013-02-27)
Nina Bloecher, Rocky de Nys, Andrew J Poole, Jana Guenther
ABSTRACT
The hydroid Ectopleura larynx is one of the main fouling organisms on salmon aquaculture cages in Norway; this study investigated novel surface materials and microtopographies to deter its settlement. The settlement preferences of hydroid larvae for 12 materials with wettabilities ranging from hydrophobic (54°) to hydrophilic (112°) were tested in a no-choice bioassay. Although settlement differed between materials, with the highest average settlement on polytetrafluoroethylene (95%) and the lowest on untreated polyurethane (53%), no trend regarding the tested wettabilities could be found and none of the tested materials was able to reduce average settlement below 50%. Furthermore, nine high-density polyethylene (HDPE, 100-600 μm microtopographies) and seven polydimethylsiloxane (PDMS; 40-400 μm microtopographies) microtextured surfaces were tested. There was no systematic effect of microtopography on the settlement of E. larynx larvae. However, there was a preference for settlement in channels on PDMS microtopographies between 80 and 300 μm. Similarly, there were no preferences for any of the examined microtopographies in a 12-day field test using PDMS surfaces at a commercial fish farm. The study indicated that neither surface wettability (hydrophilicity-phobicity) nor microtopographies were effective at deterring the settlement of the hydroid E. larynx. The high plasticity of the aboral pole and the hydrorhiza of the hydroids may explain settlement even under unfavourable conditions, highlighting the successful colonisation traits of this dominant biofouling species.
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Polyethylene (LDPE), ERM®, certified reference material