Skip to Content
Merck
All Photos(2)

Key Documents

427985

Sigma-Aldrich

Polyethylene

High density, melt index 12 g/10 min (190 °C/2.16kg)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

pellets

melt index

12 g/10 min (190 °C/2.16kg)

hardness

66 (Shore D, ASTM D 2240)

mp

125-140 °C

transition temp

softening point 125 °C (Vicat, ASTM D 1525)

density

0.952 g/mL at 25 °C

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Injection molded car seats, mower parts and pails.

Features and Benefits

Excellent stiffness and impact and warp resistance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Maria C S Inacio et al.
Acta orthopaedica, 84(2), 145-152 (2013-03-15)
There is no substantial clinical evidence for the superiority of alternative bearings in total knee arthroplasty (TKA). We compared the short-term revision risk in alternative surface bearing knees (oxidized zirconium (OZ) femoral implants or highly crosslinked polyethylene (HXLPE) inserts) with
Eliandra Mirlei Rossi et al.
Journal of infection in developing countries, 7(3), 229-234 (2013-03-16)
Contaminated sponges might lead to cross-contamination in kitchens since they can transfer microorganisms to surfaces where microorganisms can survive for hours or days and contaminate food. The main objective of this study was to evaluate the transfer and the survival
Morteza Meftah et al.
The Journal of bone and joint surgery. American volume, 95(13), 1193-1197 (2013-07-05)
Ceramic femoral heads produce less wear of the opposing polyethylene than do metal femoral heads in wear simulation studies. This is a matched-pair analysis of the wear of ceramic and metal femoral heads on conventional polyethylene in uncemented total hip
Alicia E Leadford et al.
Pediatrics, 132(1), e128-e134 (2013-06-05)
Hypothermia contributes to neonatal mortality and morbidity, especially in preterm and low birth weight infants in developing countries. Plastic bags covering the trunk and extremities of very low birth weight infants reduces hypothermia. This technique has not been studied in
Jelena Juloski et al.
The journal of adhesive dentistry, 15(2), 123-130 (2013-03-28)
To assess the shear bond strength to unground human enamel (ESBS) and flexural strength (FS) of different reinforcing fibers used in combination with a flowable composite resin. For ESBS testing, 90 human molars were selected and randomly divided into 9

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service