Merck
  • Deglycosylation as a mechanism of inducible antibiotic resistance revealed using a global relational tree for one-component regulators.
Deglycosylation as a mechanism of inducible antibiotic resistance revealed using a global relational tree for one-component regulators.
Chemistry & biology (2013-02-27)
Leslie Cuthbertson, Sang Kyun Ahn, Justin R Nodwell
ABSTRACT
The ligands that interact with the vast majority of small-molecule binding transcription factors are unknown, a significant gap in our understanding of sensory perception by cells. TetR-family regulators (TFRs) are found in most prokaryotes and are involved in regulating virtually every aspect of prokaryotic life however only a few TFRs have been characterized. We report the application of phylogenomics to the identification of cognate ligands for TFRs. Using phylogenomics we identify a TFR, KijR, that responds to the antibiotic kijanimicin. We go on to show that KijR represses a gene, kijX, which confers resistance to kijanimicin. Finally we show that KijX inactivates kijanimicin by the hydrolytic removal of sugar residues. This is a demonstration of antibiotic resistance by deglycosylation.
Product Description
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline hydrochloride, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)