Merck
  • Electrospray ionization on porous spraying tips for direct sample analysis by mass spectrometry: enhanced detection sensitivity and selectivity using hydrophobic/hydrophilic materials as spraying tips.
Electrospray ionization on porous spraying tips for direct sample analysis by mass spectrometry: enhanced detection sensitivity and selectivity using hydrophobic/hydrophilic materials as spraying tips.
Rapid communications in mass spectrometry : RCM (2013-02-19)
Melody Yee-Man Wong, Ho-Wai Tang, Sin-Heng Man, Ching-Wan Lam, Chi-Ming Che, Kwan-Ming Ng
ABSTRACT
Despite various porous materials having been widely adopted as spraying tips for direct sample analysis using electrospray ionization mass spectrometry (ESI-MS), the effect of surface property and porosity of spraying tip materials on their analytical performances is not clear. Investigation of their relationships could provide insight into the proper choice and/or design of spraying tip materials for direct sample analysis. The effect of spraying tip materials with different polarities, including polyester and polyethylene (hydrophobic) and wood (hydrophilic), on the detection sensitivity for a variety of compounds, and on the ESI onset voltage, were studied using ESI-MS. The porosity of each type of spraying tip was characterized by scanning electron microscopy (SEM). Factors governing the detection sensitivity were determined based on the correlation of the detection sensitivity to the ESI onset voltage, the polarity, and the porosity of the spraying tip materials. Hydrophobic tips (i.e., polyester and polyethylene) show better detection sensitivity for polar compounds but not for non-polar compounds, while hydrophilic tips (wooden tips) show the opposite effect. This phenomenon could be due to the difference in interaction between the analytes and the tips, causing the analytes to adsorb on the tip to different extents. In addition, the micro-porous nature of the tips could facilitate solvent diffusion for transporting analytes to the tip and maintain a stable spray for recording MS data. With the proper choice of spraying tip materials, trace amount of analytes at the picomole level can be detected with minimal sample pretreatment. Both the polarity and the porosity of the spraying tip materials could significantly affect detection sensitivity for a wide variety of analytes. With proper choice of spraying tip material, ESI on a porous spraying tip could be a sensitive method for the direct analysis of daily life samples.
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Polyethylene (LDPE), ERM®, certified reference material
Sigma-Aldrich
Protoporphyrin IX disodium salt, ≥90% (HPLC)