Merck
  • [Simultaneous determination of chlorpromazine and promethazine and their main metabolites by capillary electrophoresis with electrochemiluminescence].
[Simultaneous determination of chlorpromazine and promethazine and their main metabolites by capillary electrophoresis with electrochemiluminescence].
Se pu = Chinese journal of chromatography (2013-01-05)
Xufei Li, Yanying Yang, Kaowen Zhou
ABSTRACT
Based on the phenomenon that each of chlorpromazine (CPZ), promethazine (PMZ), chlorpromazine sulfoxide (CPZSO) and promethazine sulfoxide (PMZSO) could enhance the electrochemiluminescence (ECL) intensity of tris(2,2'-bipyridyl) ruthenium, a novel and sensitive method was proposed for the simultaneous determination of CPZ, PMZ and their main metabolites using capillary electrophoresis (CE) coupled with ECL detection. The influences of several experimental parameters were explored. The optimum experimental conditions were as follows: detection potential of 1. 20 V (Ag/AgCl), 40 mmol/L of phosphate buffer solution (pH 6.5) containing 5 mmol/L tris(2,2'-bipyridyl) ruthenium in ECL detection cell, running buffer solution of 18 mmol/L (pH 4.8), sample injection of 8 s at 11 kV, and separation voltage of 13.5 kV. The detection limits (3sigma) of this method were 8.3 x 10(-7) g/L for CPZ, 7.2 x 10(-6) g/L for PMZ, 1.9 x 10(-5) g/L for CPZSO and 3.7 x 10(-6) g/L for PMZSO. The linear ranges of ECL intensity versus mass concentration of medicaments were 7. 1 x 10(-6) - 6. 3 x 10(-3) g/L for CPZ, 7.5 x 10(-5) - 4.6 x 10(-3) g/L for PMZ, 9.7 x 10(-5) - 3.6 x 10(-3) g/L for CPZSO and 8.1 x 10(-5) - 7.7 x 10(-3) g/L for PMZSO. The relative standard deviations (RSDs) of the four target compounds were not more than 3% for ECL intensity and 1% for migration time. This method has the merits of simplicity, speediness, sensitivity, small sample injection, and free from interference. This method was successfully utilized to directly and simultaneously detect CPZ, PMZ, CPZSO and PMZSO in urine samples of pet dogs.
Product Description
Promethazine impurity D, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpromazine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Chlorpromazine impurity A, European Pharmacopoeia (EP) Reference Standard