Merck
  • Synthesis of phenanthridinones via palladium-catalyzed C(sp2)-H aminocarbonylation of unprotected o-arylanilines.
Synthesis of phenanthridinones via palladium-catalyzed C(sp2)-H aminocarbonylation of unprotected o-arylanilines.
Chemical communications (Cambridge, England) (2012-11-22)
Dongdong Liang, Ziwei Hu, Jiangling Peng, Jinbo Huang, Qiang Zhu
ABSTRACT
An efficient synthesis of free (NH)-phenanthridinones through Pd-catalyzed C(sp(2))-H aminocarbonylation of unprotected o-arylanilines under an atmospheric pressure of CO has been developed. Some ortho heteroarene substituted anilines as well as N-alkyl protected o-arylanilines are also suitable substrates for this C-H aminocarbonylation reaction.
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Supelco
Aniline, analytical standard
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Aniline hydrochloride, ≥99%