Merck
  • Effect of relative humidity on the migration of benzophenone from paperboard into the food simulant Tenax® and modelling hereof.
Effect of relative humidity on the migration of benzophenone from paperboard into the food simulant Tenax® and modelling hereof.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2012-11-17)
L L Barnkob, J H Petersen
ABSTRACT
The aim of this study was to investigate the effect of relative humidity on the migration of benzophenone from paperboard into the food simulant Tenax®. Kinetic migration investigations were carried out with three relative humidities in the interval between 39% and >73%. All investigations were carried out at a constant temperature of 34°C. It was found that the migration of benzophenone after more than 30 days was 4.8 times higher at a relative humidity of 64%-71%, and 7.3 times higher at a relative humidity of >73%, compared with that at a relative humidity of 39%-49%. Diffusion and partition coefficients were derived from the results by using a software for modelling migration in multilayer materials. Both the diffusion coefficient and the partition coefficient, between paperboard and Tenax®, decrease with increasing relative humidity. The experimental results were correctly modelled only when the paperboard was regarded as a one-layer system as compared with a two-layer system: where the main part of the paperboard (B1) has a high diffusion rate and a thin part of the paperboard (B2) in contact with the foodstuff has a lower diffusion rate.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Supelco
Melting point standard 47-49°C, analytical standard
Sigma-Aldrich
Benzophenone, SAJ first grade, ≥99.0%
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Benzophenone, European Pharmacopoeia (EP) Reference Standard