Merck
  • Characterization of a vacuolar zinc transporter OZT1 in rice (Oryza sativa L.).
Characterization of a vacuolar zinc transporter OZT1 in rice (Oryza sativa L.).
Molecular biology reports (2012-10-17)
Hong-Xia Lan, Zhou-Fei Wang, Qi-Hong Wang, Mei-Mei Wang, Yong-Mei Bao, Ji Huang, Hong-Sheng Zhang
ABSTRACT
The CDF family is a ubiquitous family that has been identified in prokaryotes, eukaryotes, and archaea. Members of this family are important heavy metal transporters that transport metal ions out of the cytoplasm. In this research, a full length cDNA named Oryza sativa Zn Transporter 1 (OZT1) that closely related to rat ZnT-2 (Zn Transporter 2) gene was isolated from rice. The OZT1 encoding a CDF family protein shares 28.2 % ~ 84.3 % of identities and 49.3 % ~ 90.9 % of similarities with other zinc transporters such as RnZnT-2, HsZnT-8, RnZnT-8 and AtMTP1. OZT1 was constitutively expressed in various rice tissues. The OZT1 expression was significantly induced both in the seedlings of japonica rice Nipponbare and indica rice IR26 in response to Zn(2+) and Cd(2+) treatments. Besides, OZT1 expression was also increased when exposed to other excess metals, such as Cu(2+), Fe(2+) and Mg(2+). Subcellular localization analysis indicated that OZT1 localized to vacuole. Heterologous expression of OZT1 in yeast increased tolerance to Zn(2+) and Cd(2+) stress but not the Mg(2+) stress. Together, OZT1 is a CDF family vacuolar zinc transporter conferring tolerance to Zn(2+) and Cd(2+) stress, which is important to transporting and homeostasis of Zn, Cd or other heavy metals in plants.
Sigma-Aldrich
Zinc sulfate heptahydrate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, meets USP testing specifications
Sigma-Aldrich
Zinc sulfate heptahydrate, BioUltra, for molecular biology, 2.0 M in H2O
Sigma-Aldrich
Zinc sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Zinc sulfate heptahydrate, suitable for plant cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Zinc sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Zinc sulfate heptahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Zinc sulfate heptahydrate, ACS reagent, 99%
Sigma-Aldrich
Zinc sulfate solution, 0.3 N
Sigma-Aldrich
Cadmium chloride, technical grade
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis