Merck
  • Social scent marks do not improve avoidance of parasites in foraging bumblebees.
Social scent marks do not improve avoidance of parasites in foraging bumblebees.
The Journal of experimental biology (2012-10-06)
Bertrand Fouks, H Michael G Lattorff
ABSTRACT
Foraging is a result of innate and acquired mechanisms, and is optimized in order to increase fitness. During foraging, an animal faces many threats, such as predation and infection. The uptake of parasites and diseases while foraging is common and an individual should be adapted to detect and avoid such threats, using cues from either the abiotic environment or the parasite. Social animals possess an additional cue to detect such contaminated food sources: information provided by conspecifics. Bumblebees avoid contaminated flowers, but the cues used by the bees to distinguish contamination remain unknown. Under controlled laboratory conditions, we tested the use of scent marks derived from other foragers in choosing between a contaminated (by Crithidia bombi) and an uncontaminated flower. As a positive control we tested the bee's choice between two flowers, one scented with geraniol and containing a highly rewarding sugar solution, and the other not scented and containing a poorer reward. The bees mainly chose the uncontaminated and the rewarding scented flowers. Scent marks did not increase the efficiency of the bumblebees in choosing the better flower. The bees from both experiments behaved similarly, showing that the main and most relevant cue used to choose the uncontaminated flower is the odour from the parasite itself. The adaptation of bumblebees to avoid flowers contaminated by C. bombi arose from the long-term host-parasite interaction between these species. This strong adaptation results in an innate behaviour of bees and a detection and aversion of the odour of contaminated flower nectar.
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
Supelco
Geraniol, analytical standard
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Geraniol, natural, ≥97%, FG