Merck
  • A rhodamine-deoxylactam based sensor for chromo-fluorogenic detection of nerve agent simulant.
A rhodamine-deoxylactam based sensor for chromo-fluorogenic detection of nerve agent simulant.
Bioorganic & medicinal chemistry letters (2012-09-22)
Zhisheng Wu, Xuanjun Wu, Yuhui Yang, Ting-bin Wen, Shoufa Han
ABSTRACT
N-(rhodamine B)-deoxylactam-5-amino-1-pentanol (dRB-APOH) was designed and prepared as the chromo-fluorogenic sensor for detection of a nerve agent simulant via analyte triggered tandem phosphorylation and opening of the intramolecular deoxylactam. The successful detection of diethyl chlorophosphate suggests the utility of rhodamine-deoxylactams as the chromo-fluorogenic signal reporting platform for design of sensors targeting reactive chemical species via various chemistries.
Sigma-Aldrich
1-Pentanol, SAJ first grade, ≥97.0%
Sigma-Aldrich
1-Pentanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1-Pentanol-1-13C, 99 atom % 13C, 99% (CP)
Sigma-Aldrich
1-Pentanol, ACS reagent, ≥99%
Sigma-Aldrich
1-Pentanol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Amyl alcohol, ≥99%, FG
Supelco
1-Pentanol, analytical standard
Sigma-Aldrich
1-Pentanol, ReagentPlus®, ≥99%