Merck
Combinatorial wetting in colour: an optofluidic nose.
Lab on a chip (2012-07-10)
Kevin P Raymond, Ian B Burgess, Mackenzie H Kinney, Marko Lončar, Joanna Aizenberg
ABSTRACT
We present a colourimetric litmus test for simple differentiation of organic liquids based on wetting, which achieves chemical specificity without a significant sacrifice in portability or ease-of-use. Chemical specificity is derived from the combination of colourimetric wetting patterns produced by liquids in an array of inverse opal films, each having a graded wettability, but using different surface groups to define that gradient.
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Xylenes, SAJ special grade, ≥80.0%
Sigma-Aldrich
Xylenes, SAJ first grade, ≥80.0%
Sigma-Aldrich
Xylenes, reagent grade
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Xylenes, histological grade
Supelco
o-Xylene, suitable for HPLC, 98%