Skip to Content
Merck
All Photos(1)

Key Documents

X1040

Sigma-Aldrich

o-Xylene

reagent grade, ≥98.0%

Synonym(s):

1,2-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1815558
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
bp:
143-145 °C (lit.)
vapor pressure:
<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

grade

reagent grade

Quality Level

vapor density

3.7 (vs air)

vapor pressure

<0.1 atm ( 21.1 °C)
16 mmHg ( 37.7 °C)
7 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

867 °F

expl. lim.

7 %

dilution

(for analytical testing)

refractive index

n20/D 1.505 (lit.)

bp

143-145 °C (lit.)

mp

-26--23 °C (lit.)

density

0.879 g/mL at 20 °C (lit.)

SMILES string

Cc1ccccc1C

InChI

1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3

InChI key

CTQNGGLPUBDAKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

O-Xylene is an aromatic hydrocarbon that is commonly used as a solvent in the pharmaceutical, printing, rubber, cleaning agent, paint thinner, varnish, synthetic fiber, and leather industries.

Application

O-Xylene can be used as a reactant to synthesize:      
  • Phthalic anhydride by oxidation using various catalysts.
  • 4-Nitro-o-xylene using fuming nitric acid and polyphosphoric acid in the presence of an acid catalyst.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier Alluin et al.
PloS one, 9(10), e111072-e111072 (2014-10-29)
While several cellular and pharmacological treatments have been evaluated following spinal cord injury (SCI) in animal models, it is increasingly recognized that approaches to address the glial scar, including the use of chondroitinase ABC (ChABC), can facilitate neuroanatomical plasticity. Moreover
Casper Bindzus Foldager et al.
Cartilage, 5(2), 123-132 (2014-04-01)
The objective of the present study was to investigate the presence and distribution of 2 basement membrane (BM) molecules, laminin and collagen type IV, in healthy and degenerative cartilage tissues. Normal and degenerated tissues were obtained from goats and humans
Maia Chanrion et al.
Nature communications, 5, 5005-5005 (2014-10-09)
Epithelial-to-mesenchymal transition-like (EMT-like) is a critical process allowing initiation of metastases during tumour progression. Here, to investigate its role in intestinal cancer, we combine computational network-based and experimental approaches to create a mouse model with high metastatic potential. Construction and
Pawaree Saisawat et al.
Kidney international, 85(6), 1310-1317 (2013-10-25)
Congenital abnormalities of the kidney and urinary tract (CAKUT) account for approximately half of children with chronic kidney disease and they are the most frequent cause of end-stage renal disease in children in the US. However, its genetic etiology remains
Tanya Aggarwal et al.
Neurobiology of aging, 35(8), 1929-1938 (2014-03-19)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disease characterized by selective loss of upper and lower motor neurons and skeletal muscle atrophy. Epidemiologic and experimental evidence suggest the involvement of androgens in ALS pathogenesis, but the mechanism through which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service