Merck
  • Anti-inflammatory effect of IMMLG5521, a coumarin derivative, on Sephadex-induced lung inflammation in rats.
Anti-inflammatory effect of IMMLG5521, a coumarin derivative, on Sephadex-induced lung inflammation in rats.
International immunopharmacology (2012-07-10)
Zhipeng Li, Jinfeng Hu, Mingna Sun, Haijie Ji, Shifeng Chu, Gang Liu, Naihong Chen
ABSTRACT
The study is to investigate the effect of compound IMMLG5521, a coumarin derivative, on lung inflammation induced by Sephadex in rats. Sephadex led to massive granulomas, infiltration of neutrophils and eosinophils, the increase of TNF-α level in BALF and lung tissue. Sephadex injection led to the up-regulation of VCAM-1 and ICAM-1 expressions in the lungs. However, pretreatment with IMMLG5521 (6.25 and 12.5 mg/kg) inhibited massive granulomas and infiltration of neutrophils and eosinophils, decreased TNF-α level in BALF and lung tissue, and down-regulated VCAM-1 and ICAM-1 expressions in the lung tissue. In conclusion, compound IMMLG5521 may suppress the lung injury induced by Sephadex, at least in part, due to the prevention of the up-regulation of VCAM-1 and ICAM-1 expressions and TNF-α level.
Sigma-Aldrich
Sephadex® G-100, BioReagent, for molecular biology, DNA grade, medium
Sigma-Aldrich
Sephadex® G-25, BioReagent, for molecular biology, DNA grade, Superfine
Sigma-Aldrich
Sephadex® G-75, Superfine
Sigma-Aldrich
Sephadex® G-75, Medium
Sigma-Aldrich
Sephadex® G-100, Medium
Sigma-Aldrich
Sephadex® G-25, Medium
Sigma-Aldrich
Sephadex® G-25, Coarse
Sigma-Aldrich
Sephadex® G-15, Medium
Sigma-Aldrich
Sephadex® G-25, Fine
Sigma-Aldrich
Sephadex® G-100, Superfine
Sigma-Aldrich
Sephadex® G-50, BioReagent, for molecular biology, DNA grade, fine
Supelco
Sephadex® G-50, Medium
Sigma-Aldrich
Sephadex® G-10, Medium
Sigma-Aldrich
Sephadex® G-50, BioReagent, for molecular biology, DNA grade, medium
Sigma-Aldrich
Sephadex® G-50, Superfine
Sigma-Aldrich
Sephadex® G-50, Fine