Merck
  • Distinguishing pancreatic cancer from chronic pancreatitis and healthy individuals by (1)H nuclear magnetic resonance-based metabonomic profiles.
Distinguishing pancreatic cancer from chronic pancreatitis and healthy individuals by (1)H nuclear magnetic resonance-based metabonomic profiles.
Clinical biochemistry (2012-05-23)
Lin Zhang, Haifeng Jin, Xuegang Guo, Zhao Yang, Lina Zhao, Shanhong Tang, Ping Mo, Kaichun Wu, Yongzhan Nie, Yanglin Pan, Daiming Fan
ABSTRACT
To develop a noninvasive and accessible diagnostic method for pancreatic cancer (PC). We presented a metabolomic method, pattern recognition techniques applied to (1)H nuclear magnetic resonance ((1)H NMR) spectra, to investigate the plasma metabolites obtained from 19 patients with PC, 20 patients with chronic pancreatitis (CP) and 20 healthy individuals. Metabolic changes associated with PC included abnormal amino acid and lipid metabolism, and possible multiple metabolic syndrome. PC elevated plasma levels of N-acetyl glycoprotein (NAG), dimethylamine (DMA), very low density lipoprotein (VLDL), and acetone, and reduced levels of 3-hydroxybutyrate, lactate, high density lipoprotein (HDL), low density lipoprotein (LDL), citrate, alanine, glutamate, glutamine, histidine, isoleucine, lysine, and valine. These metabolites could be a biomarker group for PC that distinguishes between PC and CP patients and healthy individuals. NMR-based metabonomic strategy appears as a promising approach for distinguishing pancreatic cancer and identifying new strategies for prevention or therapy in the clinical practice.
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethylamine solution, CP, 50% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethylamine hydrochloride, purum, ≥98.0% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethylamine hydrochloride, 99%
Pricing and availability is not currently available.