Merck
  • Deferoxamine pretreatment prevents Cr(VI)-induced nephrotoxicity and oxidant stress: role of Cr(VI) chelation.
Deferoxamine pretreatment prevents Cr(VI)-induced nephrotoxicity and oxidant stress: role of Cr(VI) chelation.
Toxicology (2011-11-26)
Eduardo Molina-Jijón, Guillermo Zarco-Márquez, Omar Noel Medina-Campos, Zyanya Lucía Zataraín-Barrón, Rogelio Hernández-Pando, Enrique Pinzón, Rosa Marina Zavaleta, Edilia Tapia, José Pedraza-Chaverri
ABSTRACT
Deferoxamine (DFO) is a recognized iron chelator which has been shown to exert nephroprotection in models of toxic nephropathies. In the present work the potential protective effects of DFO against Cr(VI)-induced nephrotoxicity and oxidant stress were evaluated. Rats were injected with a single injection (15mg/kg, s.c.) of potassium dichromate (K(2)Cr(2)O(7)). DFO was given as a single i.p. injection 30min before K(2)Cr(2)O(7) administration at three different doses (100, 200 and 400mg/kg). It was found that DFO pretreatment attenuated, in a dose-dependent way, K(2)Cr(2)O(7)-induced renal dysfunction and structural alterations evaluated by serum creatinine, blood urea nitrogen, creatinine clearance, proteinuria, plasma glutathione peroxidase activity, urinary excretion of N-acetyl-β-d-glucosaminidase and histological analyses. Furthermore, DFO prevented the K(2)Cr(2)O(7)-induced renal oxidant stress and the decrease in the activity of the antioxidant enzymes superoxide dismutase, glutathione reductase, glutathione peroxidase, glutathione-S-transferase and catalase. Finally it was found that DFO, at 400mg/kg, decreases renal Cr(VI) content which prompted us to evaluate the potential Cr(VI) chelating properties of this compound. Indeed was found in an in vitro assay that DFO was an effective Cr(VI) chelator with an IC(50) of 800μg. In additional groups of rats was found that DFO posttreatment was ineffective to attenuate K(2)Cr(2)O(7)-induced nephrotoxicity and renal oxidant stress. Furthermore, DFO was unable to modify urinary excretion of total chromium. The nephroprotective effect of DFO against Cr(VI)-induced nephrotoxicity and oxidant stress may be explained, at least partially, by the ability of DFO to chelate Cr(VI) and to attenuate renal Cr(VI) content. However, it cannot be excluded that the ability of DFO to chelate iron may also be involved in the protection observed in our study.
Sigma-Aldrich
Chromium standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Cr, 1000 ppm Cr
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium dichromate solution, 1/60 M
Pricing and availability is not currently available.
Supelco
Potassium dichromate, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium dichromate, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium dichromate, BioXtra, ≥99.5%
Pricing and availability is not currently available.