Merck
  • Controlled production of polymer microspheres from microgel-stabilized high internal phase emulsions.
Controlled production of polymer microspheres from microgel-stabilized high internal phase emulsions.
Chemical communications (Cambridge, England) (2010-08-24)
Zifu Li, Xiaoling Wei, To Ngai
ABSTRACT
We describe a flexible method for large-scale production of polymeric microspheres by templating a microgel-stabilized oil-in-water (o/w) high internal phase emulsion.
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Xylenes, reagent grade
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
Xylenes, SAJ special grade, ≥80.0%
Sigma-Aldrich
Xylenes, SAJ first grade, ≥80.0%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Xylenes, histological grade
Sigma-Aldrich
Xylenes, ACS reagent, ≥98.5% xylenes + ethylbenzene basis
Supelco
o-Xylene, suitable for HPLC, 98%