Merck
  • Demineralized dentin 3D porosity and pore size distribution using mercury porosimetry.
Demineralized dentin 3D porosity and pore size distribution using mercury porosimetry.
Dental materials : official publication of the Academy of Dental Materials (2009-01-29)
Elsa Vennat, Christine Bogicevic, Jean-Marie Fleureau, Michel Degrange
ABSTRACT
The objectives of this study were to assess demineralized dentin porosity and quantify the different porous features distribution within the material using mercury intrusion porosimetry (MIP) technique. We compared hexamethyldisilazane (HMDS) drying and lyophilization (LYO) (freeze-drying) in sample preparation. Fifty-six dentin discs were assigned into three groups. The control (CTR) group discs were superficially acid-etched (15s 37% H(3)PO(4)) to remove the smear layer and then freeze-dried whereas LYO and HMDS groups samples were first totally demineralized using EDTA 0.5M and then freeze-dried and HMDS-dried respectively. MIP was used to determine open porosity and pore size distribution of each pair of samples. Field emission scanning electron microscopy (FESEM) was used to illustrate the results. The results showed two types of pores corresponding either to tubules and micro-branches or to inter-fibrillar spaces created by demineralization. Global porosity varied from 59% (HMDS-dried samples) to 70% (freeze-dried samples). Lyophilization drying technique seems to lead to less shrinkage than HMDS drying. FESEM revealed that collagen fibers of demineralized lyophilized samples are less melted together than in the HMDS-dried samples. Demineralized dentin porosity is a key parameter in dentin bonding that will influence the hybrid layer quality. Its characterization could be helpful to improve the monomers infiltration.
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide, 97%
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]yttrium
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 40% in THF
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF