Skip to Content
Merck
All Photos(1)

Key Documents

80631

Sigma-Aldrich

Sodium bis(trimethylsilyl)amide solution

40% in THF

Synonym(s):

HMN-Na Sol, NaHMDS solution, Hexamethyldisilazane sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NNa
CAS Number:
Molecular Weight:
183.37
Beilstein:
3629917
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

40% in THF

SMILES string

C[Si](C)(C)N([Na])[Si](C)(C)C

InChI

1S/C6H18NSi2.Na/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

WRIKHQLVHPKCJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Sodium bis(trimethylsilyl)amide, also known as NaHMDS, is a metal dialkylamide reagent that combines both high basicity and nucleophilicity. It is a general reagent used for the enolization of carbonyl compounds.
  • It can be used to prepare phosphonium ylides in Wittig reaction.
  • NaHMDS solution can also be used as a source of nitrogen in aminomethylation and in the synthesis of primary amines from the corresponding alkyl halides.

Other Notes

prices for bulk quantities on request

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Hexamethyldisilazide.
Watson B T and Lebel H
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2005)
Regio and stereoselective preparation of enolates from ketones by means of sodium bis (trimethylsilyl)-azide.
Gaudemar M and Bellassoued M
Tetrahedron Letters, 30(21), 2779-2782 (1989)
Silylated Amines II. A New, Highly Variable Amine Synthesis by the N,N-Bis(trimethylsilyl)aminomethylation of Grignard Compounds.
Bestmann H J, et al.
Synthesis, 1987(09), 848-850 (1987)
Eine einfache Synthese primarer Amine uber ihre N, N?Bis (trimethylsilyl)?Derivate.
Bestmann H J and Wolfel G
Chemische Berichte, 117(3), 1250-1254 (1984)
(E)?Selective Wittig Reactions between a Nonstabilized Phosphonium Ylide Bearing a Phosphastibatriptycene Skeleton and Benzaldehydes.
Uchiyama Y, et al.
European Journal of Organic Chemistry, 2017(1), 159-174 (2017)

Articles

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service