Merck
  • Regular self-motion of a liquid droplet powered by the chemical marangoni effect.
Regular self-motion of a liquid droplet powered by the chemical marangoni effect.
Colloids and surfaces. B, Biointerfaces (2006-12-16)
Ken Nagai, Yutaka Sumino, Kenichi Yoshikawa
ABSTRACT
We describe here our recent work on spontaneous regular motion of liquid droplet powered by the chemical Marangoni effect under spatially symmetric conditions. It is shown that a spontaneously crawling oil droplet on a glass substrate with a nonequilibrium chemical condition of cationic surfactant exhibits regular rhythmic motion in a quasi-one-dimensional vessel, whereas irregular motion is induced in a two-dimensionally isotropic environment. Such behavior of a droplet demonstrates that spontaneous regular motion can be generated under fluctuating conditions by imposing an appropriate geometry. As another system, we introduce alcohol droplet moving spontaneously on water surface. The droplet spontaneously forms a specific morphology depending on its volume, causing specific mode of translational motion. An alcohol droplet with a smaller volume floating on water surface moves irregularly. On the other hand, a droplet with a larger volume undergoes vectorial motion accompanied by deformation into an asymmetric shape. This result suggests a scenario on the emergence of regular motion coupled with geometrical pattern formation under far-from-equilibrium conditions.
Sigma-Aldrich
1-Pentanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1-Pentanol, SAJ first grade, ≥97.0%
Sigma-Aldrich
1-Pentanol-1-13C, 99 atom % 13C, 99% (CP)
Sigma-Aldrich
Trimethyloctadecylammonium chloride, ≥95.0% (calc. on dry substance, T)
Supelco
1-Pentanol, analytical standard
Sigma-Aldrich
Amyl alcohol, ≥99%, FG
Sigma-Aldrich
1-Pentanol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
1-Pentanol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Pentanol, ACS reagent, ≥99%
Sigma-Aldrich
Trimethyloctadecylammonium bromide, 98%