Merck
  • Anodic modification of proline derivatives using a lithium perchlorate/nitromethane electrolyte solution.
Anodic modification of proline derivatives using a lithium perchlorate/nitromethane electrolyte solution.
Organic letters (2002-10-12)
Shokaku Kim, Kanako Hayashi, Yoshikazu Kitano, Masahiro Tada, Kazuhiro Chiba
ABSTRACT
[reaction: see text] N-Acyliminium cation of prolines was efficiently generated to accumulate in an undivided cell at 0 degrees C by an anodic oxidation of N-acylprolines or alpha'-phenylsulfanylated N-acylproline derivatives in a lithium perchlorate/nitromethane solution. The iminium cation intermediates gave modified prolines by a reaction with nucleophiles under mild conditions.
Sigma-Aldrich
Lithium perchlorate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, ACS reagent, ≥95.0%
Sigma-Aldrich
Lithium perchlorate, purum p.a., ≥98.0% (calc. based on dry substance, T), powder