Merck
  • Cycloaddition reaction of 2-azadienes derived from beta-amino acids with electron-rich and electron-deficient alkenes and carbonyl compounds. Synthesis of pyridine and 1,3-oxazine derivatives.
Cycloaddition reaction of 2-azadienes derived from beta-amino acids with electron-rich and electron-deficient alkenes and carbonyl compounds. Synthesis of pyridine and 1,3-oxazine derivatives.
The Journal of organic chemistry (2002-04-02)
Francisco Palacios, Esther Herrán, Gloria Rubiales, Jose María Ezpeleta
ABSTRACT
Functionalized keto-enamines 6 were obtained by nucleophilic addition of enol ethers to the imine moiety of 2-azadienes derived from dehydroaspartic esters 4. Reactions of 2-azadiene 4c containing three electron-withdrawing substituents (CO(2)R) with enol ethers 5 in the presence of lithium perchlorate led to the formation of tetrahydropyridine derivatives 7 in a regio- and stereoselective fashion. 2H-[1,3]-oxazines 10 and pyridine derivatives 12 and 13 were obtained by heterocycloaddition reactions of electron-poor azadienes 4d-g containing two electron-withdrawing substituents (4-O(2)N-C(6)H(4), CO(2)R) in positions 1 and 4 with carbonyl derivatives (ethyl glyoxalate 9a and diethyl ketomalonate 9b) and the electron-deficient olefin tetracyanoethylene 11.
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Lithium perchlorate, purum p.a., ≥98.0% (calc. based on dry substance, T), powder
Sigma-Aldrich
Lithium perchlorate, ACS reagent, ≥95.0%