Merck
  • Mitochondrial metabolism supports resistance to IDH mutant inhibitors in acute myeloid leukemia.
Mitochondrial metabolism supports resistance to IDH mutant inhibitors in acute myeloid leukemia.
The Journal of experimental medicine (2021-03-25)
Lucille Stuani, Marie Sabatier, Estelle Saland, Guillaume Cognet, Nathalie Poupin, Claudie Bosc, Florence A Castelli, Lara Gales, Evgenia Turtoi, Camille Montersino, Thomas Farge, Emeline Boet, Nicolas Broin, Clément Larrue, Natalia Baran, Madi Y Cissé, Marc Conti, Sylvain Loric, Tony Kaoma, Alexis Hucteau, Aliki Zavoriti, Ambrine Sahal, Pierre-Luc Mouchel, Mathilde Gotanègre, Cédric Cassan, Laurent Fernando, Feng Wang, Mohsen Hosseini, Emeline Chu-Van, Laurent Le Cam, Martin Carroll, Mary A Selak, Norbert Vey, Rémy Castellano, François Fenaille, Andrei Turtoi, Guillaume Cazals, Pierre Bories, Yves Gibon, Brandon Nicolay, Sébastien Ronseaux, Joseph R Marszalek, Koichi Takahashi, Courtney D DiNardo, Marina Konopleva, Véra Pancaldi, Yves Collette, Floriant Bellvert, Fabien Jourdan, Laetitia K Linares, Christian Récher, Jean-Charles Portais, Jean-Emmanuel Sarry
ABSTRACT
Mutations in IDH induce epigenetic and transcriptional reprogramming, differentiation bias, and susceptibility to mitochondrial inhibitors in cancer cells. Here, we first show that cell lines, PDXs, and patients with acute myeloid leukemia (AML) harboring an IDH mutation displayed an enhanced mitochondrial oxidative metabolism. Along with an increase in TCA cycle intermediates, this AML-specific metabolic behavior mechanistically occurred through the increase in electron transport chain complex I activity, mitochondrial respiration, and methylation-driven CEBPα-induced fatty acid β-oxidation of IDH1 mutant cells. While IDH1 mutant inhibitor reduced 2-HG oncometabolite and CEBPα methylation, it failed to reverse FAO and OxPHOS. These mitochondrial activities were maintained through the inhibition of Akt and enhanced activation of peroxisome proliferator-activated receptor-γ coactivator-1 PGC1α upon IDH1 mutant inhibitor. Accordingly, OxPHOS inhibitors improved anti-AML efficacy of IDH mutant inhibitors in vivo. This work provides a scientific rationale for combinatory mitochondrial-targeted therapies to treat IDH mutant AML patients, especially those unresponsive to or relapsing from IDH mutant inhibitors.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-CPT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Anti-SLC25A20 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Monoclonal Anti-Phosphoserine antibody produced in mouse, clone PSR-45, purified from hybridoma cell culture, buffered aqueous solution
Sigma-Aldrich
Anti-PGC-1α Mouse mAb (4C1.3), liquid, clone 4C1.3, Calbiochem®