Merck
  • Mechanism of Ca2+-Dependent Pro-Apoptotic Action of Selenium Nanoparticles, Mediated by Activation of Cx43 Hemichannels.
Mechanism of Ca2+-Dependent Pro-Apoptotic Action of Selenium Nanoparticles, Mediated by Activation of Cx43 Hemichannels.
Biology (2021-08-28)
Egor A Turovsky, Elena G Varlamova
ABSTRACT
To date, there are practically no data on the mechanisms of the selenium nanoparticles action on calcium homeostasis, intracellular signaling in cancer cells, and on the relationship of signaling pathways activated by an increase in Ca2+ in the cytosol with the induction of apoptosis, which is of great importance. The study of these mechanisms is important for understanding the cytotoxic effect of selenium nanoparticles and the role of this microelement in the regulation of carcinogenesis. The work is devoted to the study of the role of selenium nanoparticles obtained by laser ablation in the activation of the calcium signaling system and the induction of apoptosis in human glioblastoma cells (A-172 cell line). In this work, it was shown for the first time that the generation of Ca2+ signals in A-172 cells occurs in response to the application of various concentrations of selenium nanoparticles. The intracellular mechanism responsible for the generation of these Ca2+ signals has also been established. It was found that nanoparticles promote the mobilization of Ca2+ ions from the endoplasmic reticulum through the IP3-receptor. This leads to the activation of vesicular release of ATP through connexin hemichannels (Cx43) and paracrine cell activation through purinergic receptors (mainly P2Y). In addition, it was shown that the activation of this signaling pathway is accompanied by an increase in the expression of pro-apoptotic genes and the induction of apoptosis. For the first time, the role of Cx43 in the regulation of apoptosis caused by selenium nanoparticles in glioblastoma cells has been shown. It was found that inhibition of Cx43 leads to a significant suppression of the induction of apoptosis in these cells after 24 h treatment of cells with selenium nanoparticles at a concentration of 5 µg/mL.
Sigma-Aldrich
Hanks′ Balanced Salt Solution 10x, Without calcium chloride, magnesium sulfate and sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Carbenoxolone disodium salt, ≥98%
Sigma-Aldrich
Monensin sodium salt, 90-95% (TLC)
Sigma-Aldrich
U-73122 hydrate, powder
Sigma-Aldrich
Ryanodine, High Purity, Naturally occurring alkaloid that blocks the release of Ca2+ from the sarcoplasmic reticulum.
Sigma-Aldrich
Bafilomycin A1
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder
Sigma-Aldrich
MRS 2179 ammonium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Nystatin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Xestospongin C, film
Sigma-Aldrich
10Panx1 trifluoroacetate salt, ≥98% (HPLC)
Supelco
Amiloride Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material