Merck
  • Fruquintinib inhibits VEGF/VEGFR2 axis of choroidal endothelial cells and M1-type macrophages to protect against mouse laser-induced choroidal neovascularization.
Fruquintinib inhibits VEGF/VEGFR2 axis of choroidal endothelial cells and M1-type macrophages to protect against mouse laser-induced choroidal neovascularization.
Cell death & disease (2020-11-29)
Xiaojuan Liu, Aisong Guo, Yuanyuan Tu, Wendie Li, Lele Li, Wangrui Liu, Yuanyuan Ju, Yamei Zhou, Aimin Sang, Manhui Zhu
ABSTRACT
Wet age-related macular degeneration, which is characterized by choroidal neovascularization (CNV) and induces obvious vision loss. Vascular endothelial growth factor (VEGF) family member VEGF-A (also named as VEGF) and its receptor VEGFR2 contribute to the pathogenesis of CNV. Choroidal endothelial cells (CECs) secret C-C motif chemokine ligand 2 (CCL2), which attracts macrophages to CNV lesion and promotes macrophage M1 polarization. Accordingly, infiltrating macrophages secret inflammatory cytokines to promote CNV. In vivo, intravitreal injection of fruquintinib (HMPL-013), an antitumor neovascularization drug, alleviated mouse CNV formation without obvious ocular toxicity. Meanwhile, HMPL-013 inhibited VEGF/VEGFR2 binding in CECs and macrophages, as well as macrophage M1 polarization. In vitro, noncontact coculture of human choroidal vascular endothelial cells (HCVECs) and macrophages under hypoxia conditions was established. HMPL-013 downregulated VEGF/VEGFR2/phosphoinositide-3-kinase/protein kinase B (AKT)/nuclear factor kappa B pathway and CCL2 secretion in HCVECs, as well as VEGF/VEGFR2-induced macrophage M1 polarization under hypoxia condition. In addition, HMPL-013 inhibited HCEVC derived CCL2-induced macrophage migration and M1 polarization, along with macrophage M1 polarization-induced HCVECs proliferation, migration, and tube formation. Altogether, HMPL-013 alleviated CNV formation might via breaking detrimental cross talk between CECs and macrophages.
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), FITC conjugate, lyophilized powder
Millipore
ProteoPrep® Total Extraction Sample Kit
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O127:B8, purified by phenol extraction
Sigma-Aldrich
Mouse RANTES / CCL5 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse MCP-1 / CCL2 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates