Merck
  • Non-steroidal Anti-inflammatory Drugs Target TWISTED DWARF1-Regulated Actin Dynamics and Auxin Transport-Mediated Plant Development.
Non-steroidal Anti-inflammatory Drugs Target TWISTED DWARF1-Regulated Actin Dynamics and Auxin Transport-Mediated Plant Development.
Cell reports (2020-12-03)
Shutang Tan, Martin Di Donato, Matouš Glanc, Xixi Zhang, Petr Klíma, Jie Liu, Aurélien Bailly, Noel Ferro, Jan Petrášek, Markus Geisler, Jiří Friml
ABSTRACT
The widely used non-steroidal anti-inflammatory drugs (NSAIDs) are derivatives of the phytohormone salicylic acid (SA). SA is well known to regulate plant immunity and development, whereas there have been few reports focusing on the effects of NSAIDs in plants. Our studies here reveal that NSAIDs exhibit largely overlapping physiological activities to SA in the model plant Arabidopsis. NSAID treatments lead to shorter and agravitropic primary roots and inhibited lateral root organogenesis. Notably, in addition to the SA-like action, which in roots involves binding to the protein phosphatase 2A (PP2A), NSAIDs also exhibit PP2A-independent effects. Cell biological and biochemical analyses reveal that many NSAIDs bind directly to and inhibit the chaperone activity of TWISTED DWARF1, thereby regulating actin cytoskeleton dynamics and subsequent endosomal trafficking. Our findings uncover an unexpected bioactivity of human pharmaceuticals in plants and provide insights into the molecular mechanism underlying the cellular action of this class of anti-inflammatory compounds.
Sigma-Aldrich
2,3,4,5,6-Pentafluorothiophenol, 97%
Sigma-Aldrich
Ketorolac tris salt, ≥99%, crystalline
Sigma-Aldrich
Flurbiprofen, cyclooxygenase inhibitor
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Piroxicam, ≥98% (TLC)
Sigma-Aldrich
Tenoxicam, NSAID
Sigma-Aldrich
Nimesulide
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Meclofenamic acid sodium salt
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Sulindac, ≥98.0%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Phenylbutazone
Supelco
Isoxicam, analytical standard
Sigma-Aldrich
2,3,5-Triiodobenzoic acid, BioReagent, suitable for plant cell culture, ≥97% (HPLC)
Sigma-Aldrich
Diclofenac sodium salt
Supelco
Diflunisal, analytical standard
Sigma-Aldrich
Mefenamic acid
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Naptalam, PESTANAL®, analytical standard