Skip to Content
Merck
All Photos(1)

Key Documents

I1762

Supelco

Isoxicam

analytical standard

Synonym(s):

4-Hydroxy-2-methyl-N-5-methyl-3-isoxolyl-2H-1,2-benzothiazine-3-carboxamide-1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13N3O5S
CAS Number:
Molecular Weight:
335.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CN1C(C(=O)Nc2cc(C)on2)=C(O)c3ccccc3S1(=O)=O

InChI

1S/C14H13N3O5S/c1-8-7-11(16-22-8)15-14(19)12-13(18)9-5-3-4-6-10(9)23(20,21)17(12)2/h3-7,18H,1-2H3,(H,15,16,19)

InChI key

YYUAYBYLJSNDCX-UHFFFAOYSA-N

Gene Information

General description

Isoxicam is a long acting anti-inflammatory agent belonging to the oxicam group, and helps in relieving the symptoms of degenerative joint disease and rheumatoid arthritis. Its mode of action involves the ability to inhibit prostaglandin synthesis by suppressing the formation of cyclooxygenase and subsequent prostaglandin.

Application

Isoxicam may be used as an internal standard for the quantification of meloxicam in pharmaceutical preparations and as an analytical reference standard for the quantification of the analyte in biological samples using different analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Grace et al.
Current medical research and opinion, 10(9), 580-591 (1987-01-01)
Nineteen patients, aged 60 years and over, with rheumatoid arthritis participated in a clinical trial to investigate the pharmacokinetics of isoxicam (a new non-steroidal anti-inflammatory drug) in this age group. The purpose of the study was to determine if the
Oxicams: metabolic disposition in man and animals.
T F Woolf et al.
Drug metabolism reviews, 21(2), 255-276 (1989-01-01)
M Tariq
Research communications in chemical pathology and pharmacology, 60(1), 19-25 (1988-04-01)
The protective effect of some novel nonsteroidal anti-inflammatory agents has been studied on the endotoxin (lipopolysaccharide B) shock induced mortality in mice and biochemical changes in rats. All the three compounds included in this report, namely isoxicam, fentiazac and ketoprofen
Flow injection chemiluminescence determination of isoxicam based on diperiodatoargentate (III)-fluorescein system in a micellar medium
Zhao F and Si H
Journal of Luminescence, 135(1), 259-264 (2013)
F Bree et al.
Biochemical pharmacology, 38(5), 753-758 (1989-03-01)
Isoxicam binding to HSA was studied using equilibrium dialysis and fluorescence methods. It was shown that this drug binds to or near site I (warfarin or azapropazone site) and to site II (the diazepam site) as a secondary site, although

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service