Merck
  • New insights into antiviral and cytotoxic potential of quercetin and its derivatives - A biochemical perspective.
New insights into antiviral and cytotoxic potential of quercetin and its derivatives - A biochemical perspective.
Food chemistry (2020-07-28)
Enkhtaivan Gansukh, Arti Nile, Doo Hwan Kim, Jae Wook Oh, Shivraj Hariram Nile
ABSTRACT
Quercetin, a potential polyphenolic which possesses several biological effects. The influenza virus polymerase basic 2 (PB2) subunit of RNA polymerase responsible for replication, degree of virus conservation and active target site for designing specific antivirals. The quercetin derivatives downloaded from PubChem were screened using PyRX software configured with Vina Wizard, targeted on cap-binding site of the PB2 of influenza viral RNA polymerase. Among the PubChem library (total 97,585,747 compounds), 410 quercetin derivatives were screened using molecular docking (affinity: <-9.0 kcal) for their drug-likeness and in vitro cytopathic effect by Sulforhodamine B (SRB) assay. Among all quercetin derivatives, quercetin 3'-glucuronide (Q3G) showed strongest binding affinity towards cap-binding site of the PB2 subunit with -9.6 kcal of binding affinity and 0.00054 mM of Ki value, while quercetin 3'-glucuronide (Q7G) was presented highest anti-influenza activity with 2.10 ± 0.05 of IC50 on influenza A/PR/8/34 virus and non-cytotoxic effect as CC50 > 100 µg/mL.
Sigma-Aldrich
Quercitrin hydrate, ≥78%
Pricing and availability is not currently available.
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Rutin hydrate, ≥94% (HPLC), powder
Pricing and availability is not currently available.
Quercetin 7-glucoside, phyproof® Reference Substance
Pricing and availability is not currently available.
Sigma-Aldrich
Quercetin 3,4′-diglucoside, ≥85% (LC/MS-UV)
Pricing and availability is not currently available.
Rutin, phyproof® Reference Substance
Pricing and availability is not currently available.
Sigma-Aldrich
Oseltamivir phosphate, ≥98% (HPLC)
Pricing and availability is not currently available.