Merck
  • E4BP4-mediated inhibition of T follicular helper cell differentiation is compromised in autoimmune diseases.
E4BP4-mediated inhibition of T follicular helper cell differentiation is compromised in autoimmune diseases.
The Journal of clinical investigation (2020-03-20)
Zijun Wang, Ming Zhao, Jinghua Yin, Limin Liu, Longyuan Hu, Yi Huang, Aiyun Liu, Jiajun Ouyang, Xiaoli Min, Shijia Rao, Wenhui Zhou, Haijing Wu, Akihiko Yoshimura, Qianjin Lu
ABSTRACT
T follicular helper (Tfh) cells are indispensable for the formation of germinal center (GC) reactions, whereas T follicular regulatory (Tfr) cells inhibit Tfh-mediated GC responses. Aberrant activation of Tfh cells contributes substantially to the pathogenesis of autoimmune diseases, such as systemic lupus erythematosus (SLE). Nonetheless, the molecular mechanisms mitigating excessive Tfh cell differentiation are not fully understood. Herein we demonstrate that the adenovirus E4 promoter-binding protein (E4BP4) mediates a feedback loop and acts as a transcriptional brake to inhibit Tfh cell differentiation. Furthermore, we show that such an immunological mechanism is compromised in patients with SLE. Establishing mice with either conditional knockout (cKO) or knockin (cKI) of the E4bp4 gene in T cells reveals that E4BP4 strongly inhibits Tfh cell differentiation. Mechanistically, E4BP4 regulates Bcl6 transcription by recruiting the repressive epigenetic modifiers HDAC1 and EZH2. E4BP4 phosphorylation site mutants have limited capability with regard to inhibiting Tfh cell differentiation. In SLE, we detected impaired phosphorylation of E4BP4, finding that this compromised transcription factor is positively correlated with disease activity. These findings unveiled molecular mechanisms by which E4BP4 restrains Tfh cell differentiation, whose compromised function is associated with uncontrolled autoimmune reactions in SLE.
Sigma-Aldrich
Anti-CD3 Mouse mAb (UCHT1), liquid, >95% (immunoglobulin, SDS-PAGE), clone UCHT1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-CD28 Mouse mAb (ANC28.1/5D10), liquid, >95% (immunoglobulin, SDS-PAGE), clone ANC28.1/5D10