Merck
  • Acidic Compartment Size, Positioning, and Function during Myogenesis and Their Modulation by the Wnt/Beta-Catenin Pathway.
Acidic Compartment Size, Positioning, and Function during Myogenesis and Their Modulation by the Wnt/Beta-Catenin Pathway.
BioMed research international (2020-07-21)
Kayo M Bagri, Ivone A Rosa, Stephany Corrêa, Aline Yamashita, José Brito, Flavia Bloise, Manoel Luis Costa, Claudia Mermelstein
ABSTRACT
Lysosomes and acidic compartments are involved in breaking down of macromolecules, membrane recycling, and regulation of signaling pathways. Here, we analyzed the role of acidic compartments during muscle differentiation and the involvement of the Wnt/beta-catenin pathway in lysosomal function during myogenesis. Acridine orange was used to localize and quantify acidic cellular compartments in primary cultures of embryonic muscle cells from Gallus gallus. Our results show an increase in acidic compartment size and area, as well as changes in their positioning during the initial steps of myogenesis. The inhibition of lysosomal function by either the chloroquine Lys05 or the downregulation of LAMP-2 with siRNA impaired chick myogenesis, by inhibiting myoblast fusion. Two activators of the Wnt/beta-catenin pathway, BIO and Wnt3a, were able to rescue the inhibitory effects of Lys05 in myogenesis. These results suggest a new role for the Wnt/beta-catenin pathway in the regulation of acidic compartment size, positioning, and function in muscle cells.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Lys05, ≥98% (HPLC)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
BIO, ≥98% (HPLC)