Merck
  • Estradiol attenuates the TGF-β1-induced conversion of primary TAFs into myofibroblasts and inhibits collagen production and myofibroblast contraction by modulating the Smad and Rho/ROCK signaling pathways.
Estradiol attenuates the TGF-β1-induced conversion of primary TAFs into myofibroblasts and inhibits collagen production and myofibroblast contraction by modulating the Smad and Rho/ROCK signaling pathways.
International journal of molecular medicine (2015-07-17)
He-Song Jiang, Lei-Lei Zhu, Zheng Zhang, Hai Chen, Yun Chen, Yu-Tian Dai
ABSTRACT
The transformation of tunica albuginea-derived fibroblasts (TAFs) into myofibroblasts plays an important role in the pathological progress of Peyronie's disease (PD). However, no treatment which addresses this transformation is currently available. Estrogen has been shown to inhibit the progression of fibrosis in a number of fibrotic diseases. The aim of this study was to determine whether estrogen [17β‑estradiol (E2)] suppresses the diffentiation of primary rat TAFs into myofibroblasts in vitro. TAFs obtained from male Sprague‑Dawley rats were stimulated with either transforming growth factor‑β1 (TGF‑β1) or E2. Western blot analysis and immunofluorescence staining were used to assess changes in the expression levels of α‑smooth muscle actin (αSMA). The expression levels of additional proteins (GAPDH, p‑Smad2, Smad2, Smad4, RhoA, Rac1, ROCK1 and ROCK2) were also measured by western blot analysis. We used collagen gel assays to assess cell contractility. Additionally, the concentration of hydroxyproline in the TAF cell culture medium was detected using commercially available kits. We found that E2 reduced αSMA expression which was induced by TGF‑β1. E2 also suppressed the TGF‑β1‑induced increase in the concentration of hydroxyproline (a marker of collagen) in addition to suppressing the contraction of TAFs. The key processes affected by TGF‑β1 treatment included the phosphorylation of Smad2, ras homolog gene family, member A (RhoA) and Rho‑associated, coiled-coil containing protein kinase 2 (ROCK2); this increase in phosphorylation was inhibited by treatment with E2. Collectively, these results demonstrate that by modulating the activation of the TGF‑β1‑Smad and RhoA‑ROCK2 signaling pathways, E2 inhibited the transformation of TAFs into myofibroblasts, decreased the expression of collagen and suppressed the contraction of myofibroblasts in response to TGF-β1 stimulation.
Sigma-Aldrich
Anti-SMAD2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ROCK-2 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-GAPDH−Peroxidase antibody produced in mouse, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-RAC1 (Ab-71) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-SMA antibody, Rabbit monoclonal, clone SP171, recombinant, expressed in proprietary host, affinity isolated antibody
Sigma-Aldrich
Anti-ROCK-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-SMAD2 (pSer467) antibody produced in rabbit, affinity isolated antibody