Skip to Content
Merck
All Photos(4)

Documents

G9295

Sigma-Aldrich

Monoclonal Anti-GAPDH−Peroxidase antibody produced in mouse

clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-G3PD, Anti-G3PDH, Anti-Glyceraldehyde-3-phosphate Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GAPDH-71.1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

rabbit, canine, rat, hamster, monkey, mouse, turkey, bovine, mink, human, chicken

should not react with

prokaryotes

technique(s)

western blot: 1:35,000-1:70,000 using A431 total cell extract

isotype

IgM

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)
mouse ... Gapdh(14433)
rat ... Gapdh(24383)

General description

Monoclonal Anti-GAPDH (mouse IgM isotype) is derived from the hybridoma GAPDH-71.1 produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with rabbit GAPDH.
The gene glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is mapped to human chromosome 12p13. The protein localizes in the cytoplasm but can be translocated to the nucleus depending on cellular conditions.

Specificity

The antibody does not crossreact with non-vertebrate and prokaryote GAPDH.

Immunogen

rabbit GAPDH.

Application

Monoclonal Anti-GAPDH-Peroxidase antibody produced in mouse has been used in
  • immunoblotting
  • enzyme linked immunosorbent assay (ELISA)
  • immunocytochemistry

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is a tetramer containing identical chains. It catalyzes the reversible oxidative phosphorylation of glyceraldehyde-phosphate, which is a critical energy-yielding step in carbohydrate metabolism. It binds to several proteins including actin, tubulin, amyloid precursor, polyglutamine peptides, DRPLA (dentatorubral-pallidoluysian atrophy), and huntingtin. Protein kinase Cι/λbinds and phosphorylates GAPDH. Phosphorylated GAPDH associates with cytoskeletal elements and controls microtubule dynamics in the early secretory pathway. Poly(ADP-ribose) polymerase-1 (PARP1) interacts with GAPDH and thereby mediates brain damage in the presence of oxidative/nitrosative stress. GAPDH forms a part of OCA-S, the multicomponent OCT1 (octamer-motif-binding factor) coactivator complex, which is involved in the S phase-dependent histone H2B transcription. This association is responsible for linking H2B transcriptional machinery to cell cycle regulation and to the cellular metabolic state. GAPDH is also a component of the functional GAIT (interferon-γ-activated inhibitor of translation) mRNP (messenger ribonucleoprotein). GAPDH expression is dysregulated during melanoma progression.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% MIT.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muscle Lim Protein and myosin binding protein C form a complex regulating muscle differentiation
Arvanitis DA, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1864(12), 2308-2321 (2017)
Heterogeneous cadherin expression and multicellular aggregate dynamics in ovarian cancer dissemination
Klymenko Y, et al.
Neoplasia, 19(7), 549-563 (2017)
Bothrops jararaca venom metalloproteinases are essential for coagulopathy and increase plasma tissue factor levels during envenomation
Yamashita KM, et al.
PLoS Neglected Tropical Diseases, 8(5), e2814-e2814 (2014)
Efthalia Kerasioti et al.
Toxicology reports, 4, 364-372 (2017-09-30)
The aim of the present study was to investigate the effects of livestock feed supplemented with grape pomace (GP) or olive oil mill wastewater (OMW) byproducts on the enzymatic activity and protein expression of antioxidants enzymes, in liver and spleen
Jennifer L Whiting et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(30), E4328-E4337 (2016-07-13)
Filtration through the kidney eliminates toxins, manages electrolyte balance, and controls water homeostasis. Reabsorption of water from the luminal fluid of the nephron occurs through aquaporin-2 (AQP2) water pores in principal cells that line the kidney-collecting duct. This vital process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service