Merck
  • Adenovirus-mediated downregulation of the ubiquitin ligase RNF8 sensitizes bladder cancer to radiotherapy.
Adenovirus-mediated downregulation of the ubiquitin ligase RNF8 sensitizes bladder cancer to radiotherapy.
Oncotarget (2016-01-21)
Mei-Jun Zhao, Yan-Feng Song, Hai-Tao Niu, Ying-Xia Tian, Xu-Guang Yang, Kun Xie, Yu-Hong Jing, De-Gui Wang
ABSTRACT
The ubiquitin ligase RNF8 promotes the DNA damage response (DDR). We observed that the expression of RNF8 was increased in bladder cancer cells and that this change in RNF8 expression could be reversed by adenovirus-mediated shRNA treatment. Moreover, we found that RNF8 knockdown sensitized bladder cancer cells to radiotherapy, as demonstrated by reduced cell survival. Additionally, the absence of RNF8 induced a high rate of apoptosis and impaired double-strand break repair signaling after radiotherapy. Furthermore, experiments on nude mice showed that combining shRNF8 treatment with radiotherapy suppressed implanted bladder tumor growth and enhanced apoptotic cell death in vivo. Altogether, our results indicated that RNF8 might be a novel target for bladder cancer treatment.
Sigma-Aldrich
Anti-monoubiquityl Histone H2B (Lys119) Antibody, clone 7B4, clone 7B4, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-monoubiquitin H2A Antibody (Lys119), from rabbit, purified by affinity chromatography