Merck
  • Retinyl acetate-loaded nanoparticles: dermal penetration and release of the retinyl acetate.
Retinyl acetate-loaded nanoparticles: dermal penetration and release of the retinyl acetate.
International journal of pharmaceutics (2010-11-26)
Sunatda Arayachukeat, Supason P Wanichwecharungruang, Thapakorn Tree-Udom
ABSTRACT
Retinyl acetate (RA) loaded polymeric nanoparticle (NP) carriers were prepared using two different single polymers, ethyl cellulose (EC) and poly (ethylene glycol)-4-methoxycinnamoylphthaloylchitosan (PCPLC). The stability of RA to aqueous solution and UVA light was significantly improved when encapsulated with PCPLC, whilst EC encapsulation gave some improved stability in water but showed no improved photostability. Ex vivo application of free RA and the RA-loaded PCPLC NPs onto the surface of the freshly excised skin from a baby mouse indicated a significantly slower skin absorption rate for the encapsulated RA. However, 100% retention of the encapsulated RA in the skin tissue was observed after 24h. Confocal fluorescent analysis of the skin pieces applied with the RA-loaded PCPLC NPs indicated likely entry and accumulation of the PCPLC NPs and RA at the hair follicles. Release of RA from the PCPLC NP carriers was confirmed through the detection of an increasingly higher RA/PCPLC fluorescent signal ratio deeper into the dermis and away from the hair follicles.
Product Description
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
Retinyl acetate, analytical standard
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Sigma-Aldrich
Retinyl acetate, BioReagent, solid or viscous liquid, synthetic, suitable for cell culture
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g