Skip to Content
Merck
All Photos(3)

Key Documents

R4632

Sigma-Aldrich

Retinyl acetate

synthetic, crystalline solid or supercooled liquid

Synonym(s):

Retinol acetate, Vitamin A acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
Beilstein:
1915439
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058009
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

Assay

≥90% (HPLC)

form

crystalline solid or supercooled liquid

potency

2,600,000-2,940,000 I.U. per g

color

light yellow to dark yellow

mp

57-58 °C
~58 °C (lit.)

storage temp.

−20°C

SMILES string

CC1=C(/C=C/C(C)=C/C=C/C(C)=C/COC(C)=O)C(C)(C)CCC1

InChI

1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14+

InChI key

QGNJRVVDBSJHIZ-QHLGVNSISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vitamin A usually occurs in food as retinyl esters, which are the pre-formed sources of vitamin A. Retinyl acetate exists as yellow crystals and is insoluble in water. It is soluble in ethanol and oils.

Application

Retinyl acetate has been used:
  • to study its effects on cochlear development in mice
  • as a component of an internal standard to quantify antioxidants in yolk
  • as an internal standard to determine serum retinol concentration

Biochem/physiol Actions

Retinal, retinol and retinoic acid are the aldehyde, alcohol and acid forms of vitamin A. The retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain.
Retinoids also exist as retinyl esters such as retinyl propionate, retinyl acetate and retinyl palmitate. Retinyl esters provide pools of vitamin A that are converted into retinol and other retinoids as needed. Retinyl acetate is used in a wide range of biological applications.

Physical form

Packaged as a melt and may exist as a crystalline solid or a supercooled viscous liquid

Analysis Note

Activity based on HPLC comparison to USP standard.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capillary electrophoresis Food Additives
Frazier RA
Encyclopedia of Separation Science (2007)
The common marmoset in captivity and biomedical research
Marini RP
Common Fragrance and Flavor Materials (2018)
Effects of retinoid treatment on cochlear development, connexin expression and hearing thresholds in mice
Kim Yy, et al.
Journal of Otolaryngology - Head & Neck Surgery = Le Journal D'Oto-rhino-laryngologie et de Chirurgie Cervico-Faciale (2018)
Assessment of the relative dose-response test based on serum retinol-binding protein instead of serum retinol in determining low hepatic vitamin A stores
Fujita M, et al.
American Journal of Clinical Nutrition (2009)
Buhler Volker
Vademecum for Vitamin Formulations (2001)

Protocols

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

GB method with Ascentis® Express C30 column achieves baseline resolution for Vitamin A and E critical pair.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service